1-Boc-azetidine-3-carboxaldehyde, 97%
1-Boc-azetidine-3-carboxaldehyde, 97%
1-Boc-azetidine-3-carboxaldehyde, 97%
Thermo Scientific Chemicals

1-Boc-azetidine-3-carboxaldehyde, 97%

CAS: 177947-96-5 | C9H15NO3 | 185.223 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H52794.06
also known as H52794-06
Price (USD)
639.65
Online Exclusive
710.00
Save 70.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
639.65
Online Exclusive
710.00
Save 70.35 (10%)
Each
Chemical Identifiers
CAS177947-96-5
IUPAC Nametert-butyl 3-formylazetidine-1-carboxylate
Molecular FormulaC9H15NO3
InChI KeyJVQOZRRUGOADSU-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)N1CC(C1)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (GC)≥96.0%
FormLiquid
Free acid (titration)<2.0%
Refractive Index1.4590-1.4665 @ 20°C
1-Boc-azetidine-3-carboxaldehyde is used as a starting material in organic synthesis. It is also used as an intermediate in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-azetidine-3-carboxaldehyde is used as a starting material in organic synthesis. It is also used as an intermediate in pharmaceuticals.

Notes
Air sensitive. Store in a cool place. Incompatible with water and oxidizing agents.
RUO – Research Use Only
Li, D. B.; Rogers-Evans, M.; Carreira, E. M. Synthesis of Novel Azaspiro[3.4]octanes as Multifunctional Modules in Drug Discovery. Org. Lett. 2011, 13 (22), 6134-6136.