(S)-1-(Boc-amino)-2-propanol, 97%
(S)-1-(Boc-amino)-2-propanol, 97%
(S)-1-(Boc-amino)-2-propanol, 97%
Thermo Scientific Chemicals

(S)-1-(Boc-amino)-2-propanol, 97%

CAS: 167938-56-9 | C8H17NO3 | 175.228 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H52826.06
also known as H52826-06
Price (USD)
66.40
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
66.40
Each
Chemical Identifiers
CAS167938-56-9
IUPAC Nametert-butyl N-(2-hydroxypropyl)carbamate
Molecular FormulaC8H17NO3
InChI KeyYNJCFDAODGKHAV-UHFFFAOYNA-N
SMILESCC(O)CNC(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
FormLiquid
Assay (unspecified)>96.0%
Water Content (Karl Fischer Titration)<0.50%
Optical purity>98.0%
View more
(S)-1-(Boc-amino)-2-propanol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-1-(Boc-amino)-2-propanol is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only