DL-Cysteine, 96%
DL-Cysteine, 96%
DL-Cysteine, 96%
Thermo Scientific Chemicals

DL-Cysteine, 96%

CAS: 3374-22-9 | C3H7NO2S | 121.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H56126.06
also known as H56126-06
Price (USD)
43.65
Online Exclusive
47.90
Save 4.25 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
43.65
Online Exclusive
47.90
Save 4.25 (9%)
Each
Chemical Identifiers
CAS3374-22-9
IUPAC Name2-amino-3-sulfanylpropanoic acid
Molecular FormulaC3H7NO2S
InChI KeyXUJNEKJLAYXESH-UHFFFAOYNA-N
SMILESNC(CS)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)96%
DL-Cysteine is a NMDA agonist at quisqualate receptors at high concentration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Cysteine is a NMDA agonist at quisqualate receptors at high concentration.

Solubility
Soluble in water (partly), 1 N HCl (25 mg/ml), ethanol, and acetic acid. Insoluble in acetone.

Notes
Store at 4°C. Incompatible with oxidizing agents. Keep container tightly closed.
RUO – Research Use Only

General References:

  1. Vasil S. Minkov.; Alexander S. Krylov.; Elena V. Boldyreva.; Sergei V. Goryainov.; Sergei N. Bizyaev.; Alexander N. Vtyurin. Pressure-Induced Phase Transitions in Crystalline l- and dl-Cysteine.J. Phys. Chem. B. 2008, 112 (30),8851-8854 .
  2. James Darkwa.; Claudius Mundoma.; Reuben H. Simoyi. Antioxidant chemistry Reactivity and oxidation of DL-cysteine by some common oxidants.J. Chem. Soc., Faraday Trans. 1998, 94 (14),1971-1978 .