D-beta-Proline ethyl ester hydrochloride, 97%, Thermo Scientific Chemicals
D-beta-Proline ethyl ester hydrochloride, 97%, Thermo Scientific Chemicals
D-beta-Proline ethyl ester hydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D-beta-Proline ethyl ester hydrochloride, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H57789.03
also known as H57789-03
1 g
Catalog number H57789.03
also known as H57789-03
Price (USD)
144.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
144.00
Each
Specifications
CAS1807350-90-8
Chemical Name or MaterialD-beta-Proline ethyl ester hydrochloride
Formula Weight179.65
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Notes
Protect from heat. Store away from oxidizing agents. Refrigerate. Incompatible with heat and oxidizing agents.
RUO – Research Use Only