1-Boc-3-(methylamino)azetidine, 95%, Thermo Scientific Chemicals
1-Boc-3-(methylamino)azetidine, 95%, Thermo Scientific Chemicals
1-Boc-3-(methylamino)azetidine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Boc-3-(methylamino)azetidine, 95%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H57951.03
also known as H57951-03
1 g
Catalog number H57951.03
also known as H57951-03
Price (USD)
179.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
179.00
Each
Chemical Identifiers
CAS454703-20-9
IUPAC Nametert-butyl 3-(methylamino)azetidine-1-carboxylate
Molecular FormulaC9H18N2O2
InChI KeyCHRBSEYIEDTNSC-UHFFFAOYSA-N
SMILESCNC1CN(C1)C(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Proton NMRConforms to structure
Assay from Supplier's CofA≥94.0%
FormLiquid
1-Boc-3-(methylamino)azetidine is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-3-(methylamino)azetidine is used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Store under dry inert gas. Store away from air. Incompatible with oxidizing agents.
RUO – Research Use Only