L-Phenylalanine tert-butyl ester hydrochloride, 98%
L-Phenylalanine tert-butyl ester hydrochloride, 98%
L-Phenylalanine tert-butyl ester hydrochloride, 98%
Thermo Scientific Chemicals

L-Phenylalanine tert-butyl ester hydrochloride, 98%

CAS: 15100-75-1 | C13H20ClNO2 | 257.758 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H59122.03
also known as H59122-03
Price (USD)
51.65
Special offer
Online exclusive
Ends: 30-Jun-2026
60.90
Save 9.25 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
51.65
Special offer
Online exclusive
Ends: 30-Jun-2026
60.90
Save 9.25 (15%)
Each
Chemical Identifiers
CAS15100-75-1
IUPAC Namehydrogen tert-butyl 2-amino-3-phenylpropanoate chloride
Molecular FormulaC13H20ClNO2
InChI KeyFDMCEXDXULPJPG-UHFFFAOYNA-N
SMILES[H+].[Cl-].CC(C)(C)OC(=O)C(N)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and food industry. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and food industry. 

Solubility
Soluble in methanol.

Notes
Hygroscopic. Keep away from heat and humidity. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Martin J. O'Donnell.; Robin L. Polt. A mild and efficient route to Schiff base derivatives of amino acids. J. Org. Chem. 1982, 47 (13), 2663-2666.
  2. Takayuki Shioiri.; Kunihiro Ninomiya.; Shunichi Yamada. Diphenylphosphoryl azide. New convenient reagent for a modified Curtius reaction and for peptide synthesis. J. Am. Chem. Soc. 1972, 94 (17),6203-6205.