L-Alanine tert-butyl ester hydrochloride, 98%, Thermo Scientific Chemicals
L-Alanine tert-butyl ester hydrochloride, 98%, Thermo Scientific Chemicals
L-Alanine tert-butyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

L-Alanine tert-butyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H59359.06
also known as H59359-06
5 g
Catalog number H59359.06
also known as H59359-06
Price (USD)
387.65
Online Exclusive
431.00
Save 43.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
387.65
Online Exclusive
431.00
Save 43.35 (10%)
Each
Chemical Identifiers
CAS13404-22-3
IUPAC Nametert-butyl (2S)-2-aminopropanoate hydrochloride
Molecular FormulaC7H16ClNO2
InChI KeyWIQIWPPQGWGVHD-JEDNCBNOSA-N
SMILESCl.C[C@H](N)C(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Proton NMR>97.5%
It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Hygroscopic. Keep away from heat and humidity. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. John C. Sheehan.; Stephen L. Ledis. Total synthesis of a monocyclic peptide lactone antibiotic, etamycin.J. Am. Chem. Soc. 1973, 95 (3),875-879.
  2. James S. Nowick.; Noel A. Powell.; Tram M. Nguyen.; Glenn Noronha. An improved method for the synthesis of enantiomerically pure amino acid ester isocyanates. J. Org. Chem. 1992, 57 (26),7364-7366.