3-(2-Aminoethyl)-1-Boc-azetidine, 95%
3-(2-Aminoethyl)-1-Boc-azetidine, 95%
3-(2-Aminoethyl)-1-Boc-azetidine, 95%
Thermo Scientific Chemicals

3-(2-Aminoethyl)-1-Boc-azetidine, 95%

CAS: 898271-20-0 | C10H20N2O2 | 200.282 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H60469.03
also known as H60469-03
Price (USD)
418.65
Special offer
Online exclusive
Ends: 30-Jun-2026
492.00
Save 73.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
418.65
Special offer
Online exclusive
Ends: 30-Jun-2026
492.00
Save 73.35 (15%)
Each
Specifications
CAS898271-20-0
Chemical Name or Material3-(2-Aminoethyl)-1-Boc-azetidine
DOT InformationHazard Class: 8; Packaging Group: III
Formula Weight200.28
Health Hazard 1H314-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only