Nalpha-Boc-D-asparagine, 95%
Nalpha-Boc-D-asparagine, 95%
Nalpha-Boc-D-asparagine, 95%
Thermo Scientific Chemicals

Nalpha-Boc-D-asparagine, 95%

CAS: 75647-01-7 | C9H16N2O5 | 232.236 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H61121.14
also known as H61121-14
Price (USD)
149.65
Online Exclusive
166.00
Save 16.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
149.65
Online Exclusive
166.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS75647-01-7
IUPAC Name(2R)-2-{[(tert-butoxy)carbonyl]amino}-3-carbamoylpropanoate
Molecular FormulaC9H15N2O5
InChI KeyFYYSQDHBALBGHX-RXMQYKEDSA-M
SMILESCC(C)(C)OC(=O)N[C@H](CC(N)=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only