trans-4-(Boc-amino)cyclohexaneacetic acid, 97%
trans-4-(Boc-amino)cyclohexaneacetic acid, 97%
trans-4-(Boc-amino)cyclohexaneacetic acid, 97%
Thermo Scientific Chemicals

trans-4-(Boc-amino)cyclohexaneacetic acid, 97%

CAS: 189153-10-4 | C13H23NO4 | 257.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H62058.06
also known as H62058-06
Price (USD)
400.65
Online Exclusive
445.00
Save 44.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
400.65
Online Exclusive
445.00
Save 44.35 (10%)
Each
Specifications
CAS189153-10-4
Chemical Name or Materialtrans-4-(Boc-amino)cyclohexaneacetic acid
Formula Weight257.32
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
trans-4-(Boc-amino)cyclohexaneacetic acid used as a intermediate in pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-(Boc-amino)cyclohexaneacetic acid used as a intermediate in pharmaceutical.

Solubility
Slightly soluble in water (1.1 g/L) (25°C)

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only