L-Tyrosine methyl ester, 98%
L-Tyrosine methyl ester, 98%
L-Tyrosine methyl ester, 98%
Thermo Scientific Chemicals

L-Tyrosine methyl ester, 98%

CAS: 1080-06-4 | C10H13NO3 | 195.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number H62069.14
also known as H62069-14
Price (USD)
60.65
Special offer
Online exclusive
Ends: 30-Jun-2026
71.00
Save 10.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
60.65
Special offer
Online exclusive
Ends: 30-Jun-2026
71.00
Save 10.35 (15%)
Each
Chemical Identifiers
CAS1080-06-4
IUPAC Namemethyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
Molecular FormulaC10H13NO3
InChI KeyMWZPENIJLUWBSY-VIFPVBQESA-N
SMILESCOC(=O)[C@@H](N)CC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
L-Tyrosine methyl ester is used as a dehydroepiandrosterone as a sulfate acceptor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Tyrosine methyl ester is used as a dehydroepiandrosterone as a sulfate acceptor

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Gerald E. Clement.; James Rooney.; David Zakheim.; James Eastman. pH dependence of the dephosphorylated pepsin-catalyzed hydrolysis of N-acetyl-L-phenylalanyl-L-tyrosine methyl ester. J. Am. Chem. Soc. 1970, 92, (1), 186-189.
  2. Roger L. Peterson.; Konrad W. Hubele.; Carl Niemann. The α-Chymotrypsin-catalyzed Hydrolysis of α-N and O-Alkyl Derivatives of α-N-Acetyl-L-tyrosine Methyl Ester. Biochemistry. 1963, 2, (5), 942-946.