trans-1-(Boc-amino)-4-(2-hydroxyethyl)cyclohexane, 97%, Thermo Scientific Chemicals
trans-1-(Boc-amino)-4-(2-hydroxyethyl)cyclohexane, 97%, Thermo Scientific Chemicals
trans-1-(Boc-amino)-4-(2-hydroxyethyl)cyclohexane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-1-(Boc-amino)-4-(2-hydroxyethyl)cyclohexane, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H62087.03
also known as H62087-03
1 g
Catalog number H62087.03
also known as H62087-03
Price (USD)
377.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
377.00
Each
Specifications
CAS917342-29-1
Chemical Name or Materialtrans-1-(Boc-amino)-4-(2-hydroxyethyl)cyclohexane
Formula Weight243.34
IUPAC Nametert-butyl N-[4-(2-hydroxyethyl)cyclohexyl]carbamate
InChI KeyKTNFSGIXLVLQNK-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only