Specifications
CAS847417-37-2
Chemical Name or Materialcis-4-(Boc-amino)cyclohexaneacetaldehyde
Formula Weight241.32
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more