N-(Benzyloxycarbonyl)sarcosine, 95%
N-(Benzyloxycarbonyl)sarcosine, 95%
N-(Benzyloxycarbonyl)sarcosine, 95%
Thermo Scientific Chemicals

N-(Benzyloxycarbonyl)sarcosine, 95%

CAS: 39608-31-6 | C11H13NO4 | 223.228 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H62135.06
also known as H62135-06
Price (USD)
43.60
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
43.60
Each
Chemical Identifiers
CAS39608-31-6
IUPAC Name2-{[(benzyloxy)carbonyl](methyl)amino}acetic acid
Molecular FormulaC11H13NO4
InChI KeyCBWFTZNMONHKNZ-UHFFFAOYSA-N
SMILESCN(CC(O)=O)C(=O)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only