L-Valine benzyl ester hydrochloride, 95%
L-Valine benzyl ester hydrochloride, 95%
L-Valine benzyl ester hydrochloride, 95%
Thermo Scientific Chemicals

L-Valine benzyl ester hydrochloride, 95%

CAS: 2462-34-2 | C12H18ClNO2 | 243.73 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H62242.06
also known as H62242-06
Price (USD)
52.65
Online exclusive
57.90
Save 5.25
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
52.65
Online exclusive
57.90
Save 5.25
Each
Chemical Identifiers
CAS2462-34-2
IUPAC Namebenzyl (2S)-2-amino-3-methylbutanoate hydrochloride
Molecular FormulaC12H18ClNO2
InChI KeyZIUNABFUHGBCMF-MERQFXBCSA-N
SMILESCl.CC(C)[C@H](N)C(=O)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only