Ndelta-Benzyloxycarbonyl-L-ornithine, 98%
Ndelta-Benzyloxycarbonyl-L-ornithine, 98%
Ndelta-Benzyloxycarbonyl-L-ornithine, 98%
Thermo Scientific Chemicals

Ndelta-Benzyloxycarbonyl-L-ornithine, 98%

CAS: 3304-51-6 | C13H18N2O4 | 266.297 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H62435.06
also known as H62435-06
Price (USD)
49.65
Special offer
Online exclusive
Ends: 30-Jun-2026
58.20
Save 8.55 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
49.65
Special offer
Online exclusive
Ends: 30-Jun-2026
58.20
Save 8.55 (15%)
Each
Chemical Identifiers
CAS3304-51-6
IUPAC Name2-amino-5-{[(benzyloxy)carbonyl]amino}pentanoic acid
Molecular FormulaC13H18N2O4
InChI KeyVULSXQYFUHKBAN-UHFFFAOYNA-N
SMILESNC(CCCNC(=O)OCC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only