Nalpha-Boc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulfonyl)-L-arginine, 98%, Thermo Scientific Chemicals
Nalpha-Boc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulfonyl)-L-arginine, 98%, Thermo Scientific Chemicals
Nalpha-Boc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulfonyl)-L-arginine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nalpha-Boc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulfonyl)-L-arginine, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H62515.06
also known as H62515-06
5 g
Catalog number H62515.06
also known as H62515-06
Price (USD)
90.65
Online Exclusive
100.00
Save 9.35 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
90.65
Online Exclusive
100.00
Save 9.35 (9%)
Each
Chemical Identifiers
CAS102185-38-6
IUPAC Name5-{[amino(4-methoxy-2,3,6-trimethylbenzenesulfonamido)methylidene]amino}-2-{[(tert-butoxy)carbonyl]amino}pentanoic acid
Molecular FormulaC21H34N4O7S
InChI KeyCXZHJRGYWGPJSD-UHFFFAOYNA-N
SMILESCOC1=C(C)C(C)=C(C(C)=C1)S(=O)(=O)NC(N)=NCCCC(NC(=O)OC(C)(C)C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only