N-Boc-L-histidine methyl ester, 95%
N-Boc-L-histidine methyl ester, 95%
N-Boc-L-histidine methyl ester, 95%
Thermo Scientific Chemicals

N-Boc-L-histidine methyl ester, 95%

CAS: 2488-14-4 | C12H19N3O4 | 269.301 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H62545.03
also known as H62545-03
Price (USD)
78.65
Online Exclusive
87.50
Save 8.85 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
78.65
Online Exclusive
87.50
Save 8.85 (10%)
Each
Chemical Identifiers
CAS2488-14-4
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only