N-Fmoc-O-benzylphospho-L-threonine, 97%
N-Fmoc-O-benzylphospho-L-threonine, 97%
N-Fmoc-O-benzylphospho-L-threonine, 97%
Thermo Scientific Chemicals

N-Fmoc-O-benzylphospho-L-threonine, 97%

CAS: 175291-56-2 | C26H26NO8P | 511.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H63006.MD
also known as H63006-MD
Price (USD)
94.10
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
94.10
Each
Chemical Identifiers
CAS175291-56-2
IUPAC Name(2S,3R)-3-{[(benzyloxy)(hydroxy)phosphoryl]oxy}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Molecular FormulaC26H26NO8P
InChI KeyHOFDVXHILSPFNS-OSPHWJPCSA-N
SMILESC[C@@H](OP(O)(=O)OCC1=CC=CC=C1)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only