N-Benzyloxycarbonyl-L-tyrosine methyl ester, 98%
N-Benzyloxycarbonyl-L-tyrosine methyl ester, 98%
N-Benzyloxycarbonyl-L-tyrosine methyl ester, 98%
Thermo Scientific Chemicals

N-Benzyloxycarbonyl-L-tyrosine methyl ester, 98%

CAS: 13512-31-7 | C18H19NO5 | 329.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number H63113.14
also known as H63113-14
Price (USD)
163.65
Online Exclusive
182.00
Save 18.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
163.65
Online Exclusive
182.00
Save 18.35 (10%)
Each
Chemical Identifiers
CAS13512-31-7
IUPAC Namemethyl (2S)-2-{[(benzyloxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoate
Molecular FormulaC18H19NO5
InChI KeySLLMDHBKALJDBW-XISACWJONA-N
SMILESCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormSolid
Assay from Supplier's CofA≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only