2-Bromo-L-phenylalanine, 95%
2-Bromo-L-phenylalanine, 95%
2-Bromo-L-phenylalanine, 95%
Thermo Scientific Chemicals

2-Bromo-L-phenylalanine, 95%

CAS: 42538-40-9 | C9H10BrNO2 | 244.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63141.06
also known as H63141-06
Price (USD)
296.65
Special offer
Online exclusive
Ends: 30-Jun-2026
349.00
Save 52.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
296.65
Special offer
Online exclusive
Ends: 30-Jun-2026
349.00
Save 52.35 (15%)
Each
Chemical Identifiers
CAS42538-40-9
IUPAC Name(2S)-2-amino-3-(2-bromophenyl)propanoic acid
Molecular FormulaC9H10BrNO2
InChI KeyJFVLNTLXEZDFHW-QMMMGPOBSA-N
SMILESN[C@@H](CC1=CC=CC=C1Br)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only