N-Benzyloxycarbonyl-D-methionine, 98%
N-Benzyloxycarbonyl-D-methionine, 98%
N-Benzyloxycarbonyl-D-methionine, 98%
Thermo Scientific Chemicals

N-Benzyloxycarbonyl-D-methionine, 98%

CAS: 28862-80-8 | C13H17NO4S | 283.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H63152.06
also known as H63152-06
Price (USD)
33.10
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
33.10
Each
Chemical Identifiers
CAS28862-80-8
IUPAC Name(2R)-2-{[(benzyloxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid
Molecular FormulaC13H17NO4S
InChI KeyFPKHNNQXKZMOJJ-LLVKDONJSA-N
SMILESCSCC[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only