3-(2-Thienyl)-L-alanine, 95%
3-(2-Thienyl)-L-alanine, 95%
3-(2-Thienyl)-L-alanine, 95%
Thermo Scientific Chemicals

3-(2-Thienyl)-L-alanine, 95%

CAS: 22951-96-8 | C7H9NO2S | 171.214 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63316.06
also known as H63316-06
Price (USD)
594.65
Online Exclusive
660.00
Save 65.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
594.65
Online Exclusive
660.00
Save 65.35 (10%)
Each
Chemical Identifiers
CAS22951-96-8
IUPAC Name(2S)-2-amino-3-(thiophen-2-yl)propanoic acid
Molecular FormulaC7H9NO2S
InChI KeyWTOFYLAWDLQMBZ-LURJTMIESA-N
SMILESN[C@@H](CC1=CC=CS1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
3-(2-Thienyl)-L-alanine can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(2-Thienyl)-L-alanine can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Ole Andreas Andersen; Torgeir Flatmark; Edward Hough. Crystal Structure of the Ternary Complex of the Catalytic Domain of Human Phenylalanine Hydroxylase with Tetrahydrobiopterin and 3-(2-Thienyl)-l-alanine, and its Implications for the Mechanism of Catalysis and Substrate Activation. Journal of Molecular Biology. 2002, 320 (5), 1095-1108.
  2. Ole Andreas Andersen; Anne J Stokka; Torgeir Flatmark; Edward Hough. 2.0 Å Resolution Crystal Structures of the Ternary Complexes of Human Phenylalanine Hydroxylase Catalytic Domain with Tetrahydrobiopterin and 3-(2-Thienyl)-l-alanine or l-Norleucine: Substrate Specificity and Molecular Motions Related to Substrate Binding. Journal of Molecular Biology. 2003, 333 (4), 747-757.