4-[2-(Boc-amino)ethyl]aniline, 97%
4-[2-(Boc-amino)ethyl]aniline, 97%
4-[2-(Boc-amino)ethyl]aniline, 97%
Thermo Scientific Chemicals

4-[2-(Boc-amino)ethyl]aniline, 97%

CAS: 94838-59-2 | C13H20N2O2 | 236.315 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63324.06
also known as H63324-06
Price (USD)
346.65
Special offer
Online exclusive
Ends: 30-Jun-2026
408.00
Save 61.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
346.65
Special offer
Online exclusive
Ends: 30-Jun-2026
408.00
Save 61.35 (15%)
Each
Specifications
CAS94838-59-2
Chemical Name or Material4-[2-(Boc-amino)ethyl]aniline
ColorBrown
DOT InformationTransport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Formula Weight236.31
View more
4-[2-(Boc-amino)ethyl]aniline is used as a intermediate for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-[2-(Boc-amino)ethyl]aniline is used as a intermediate for chemical research.

Solubility
Slightly soluble in water.

Notes
Refrigerate -20°C. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only