Ndelta-Allyloxycarbonyl-Nalpha-Fmoc-L-ornithine, 95%
Ndelta-Allyloxycarbonyl-Nalpha-Fmoc-L-ornithine, 95%
Ndelta-Allyloxycarbonyl-Nalpha-Fmoc-L-ornithine, 95%
Thermo Scientific Chemicals

Ndelta-Allyloxycarbonyl-Nalpha-Fmoc-L-ornithine, 95%

CAS: 147290-11-7 | C24H26N2O6 | 438.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H63331.06
also known as H63331-06
Price (USD)
119.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
119.00
Each
Chemical Identifiers
CAS147290-11-7
IUPAC Name2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-{[(prop-2-en-1-yloxy)carbonyl]amino}pentanoic acid
Molecular FormulaC24H26N2O6
InChI KeyRXLIOYNXBHZZBI-UHFFFAOYNA-N
SMILESOC(=O)C(CCCNC(=O)OCC=C)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only