D-Alaninamide hydrochloride, 98%, Thermo Scientific Chemicals
D-Alaninamide hydrochloride, 98%, Thermo Scientific Chemicals
D-Alaninamide hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D-Alaninamide hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H63535.06
also known as H63535-06
5 g
Catalog number H63535.06
also known as H63535-06
Price (USD)
130.65
Online Exclusive
145.00
Save 14.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
130.65
Online Exclusive
145.00
Save 14.35 (10%)
Each
Chemical Identifiers
CAS71810-97-4
IUPAC Name2-aminopropanamide hydrochloride
Molecular FormulaC3H9ClN2O
InChI KeyFIAINKIUSZGVGX-UHFFFAOYNA-N
SMILESCl.CC(N)C(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
D-Alaninamide hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Alaninamide hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

Solubility
Soluble in 1% acetic acid and water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Hideo Kise; Tohru Nagashima. Enantiospecificity of α-Chymotrypsin at S'1 Site for Peptide Synthesis in Aqueous-Organic Media. Bulletin of the Chemical Society of Japan. 1993, 66 (12), 3693-3698.
  2. Mats Reslow; Patrick Adlercreutz; and Bo Mattiasson.The influence of water on protease-catalyzed peptide synthesis in acetonitrile/water mixtures. European Journal of Biochemistry. 1988, 177 (2), 313-318.