Thermo Scientific Chemicals

4-Cyano-L-phenylalanine, 95%, Thermo Scientific Chemicals

Catalog number: H63572.03
1 g, Each
Thermo Scientific Chemicals

4-Cyano-L-phenylalanine, 95%, Thermo Scientific Chemicals

Catalog number: H63572.03
1 g, Each
Quantity
Catalog number: H63572.03
also known as H63572-03
Price (USD)
Price: 135.00
Online price: 117.65
Your price:
Quantity
-

Chemical Identifiers

CAS
167479-78-9
Form
Solid
Appearance (Color)
White to off-white
Assay by NMR
>94.0%

Description

4-Cyano-L-phenylalanine used as a intermediate in organic synthesis, pharmaceutical, chemical research and biochemical application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyano-L-phenylalanine used as a intermediate in organic synthesis, pharmaceutical, chemical research and biochemical application.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text