Thermo Scientific Chemicals

4-Chloro-L-phenylalanine, 97%, Thermo Scientific Chemicals

Catalog number: H63573.MD
250 mg, Each
Thermo Scientific Chemicals

4-Chloro-L-phenylalanine, 97%, Thermo Scientific Chemicals

Catalog number: H63573.MD
250 mg, Each
Quantity
Catalog number: H63573.MD
also known as H63573-MD
Price (USD)
Price: 59.30
Online price: 50.65
Your price:
Quantity
-

Chemical Identifiers

CAS
14173-39-8
IUPAC Name
2-amino-3-(4-chlorophenyl)propanoic acid
Molecular Formula
C9H10ClNO2
InChI Key
NIGWMJHCCYYCSF-UHFFFAOYNA-N
SMILES
NC(CC1=CC=C(Cl)C=C1)C(O)=O
Assay (HPLC)
>96.0%

Description

4-Chloro-L-phenylalanine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-L-phenylalanine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

Solubility
Soluble in acetic acid. Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text