N-Fmoc-L-aspartic acid 4-methyl ester, 97%
N-Fmoc-L-aspartic acid 4-methyl ester, 97%
N-Fmoc-L-aspartic acid 4-methyl ester, 97%
Thermo Scientific Chemicals

N-Fmoc-L-aspartic acid 4-methyl ester, 97%

CAS: 145038-53-5 | C20H19NO6 | 369.373 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63774.03
also known as H63774-03
Price (USD)
34.65
Special offer
Online exclusive
Ends: 30-Jun-2026
40.10
Save 5.45 (14%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
34.65
Special offer
Online exclusive
Ends: 30-Jun-2026
40.10
Save 5.45 (14%)
Each
Chemical Identifiers
CAS145038-53-5
IUPAC Name2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-methoxy-4-oxobutanoic acid
Molecular FormulaC20H19NO6
InChI KeyHSOKCYGOTGVDHL-UHFFFAOYNA-N
SMILESCOC(=O)CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder or crystalline powder or solid
Assay by NMR≥96.0%
Proton NMRConforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only