3-[2-(Boc-amino)ethyl]azetidine, 95%
3-[2-(Boc-amino)ethyl]azetidine, 95%
3-[2-(Boc-amino)ethyl]azetidine, 95%
Thermo Scientific Chemicals

3-[2-(Boc-amino)ethyl]azetidine, 95%

CAS: 162696-31-3 | C10H20N2O2 | 200.282 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H63797.03
also known as H63797-03
Price (USD)
587.65
Special offer
Online exclusive
Ends: 30-Jun-2026
691.00
Save 103.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
587.65
Special offer
Online exclusive
Ends: 30-Jun-2026
691.00
Save 103.35 (15%)
Each
Specifications
CAS162696-31-3
Chemical Name or Material3-[2-(Boc-amino)ethyl]azetidine
ColorBrown to White
Formula Weight200.28
Health Hazard 1H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only