(R)-4-Boc-morpholine-3-carboxylic acid, 97%
(R)-4-Boc-morpholine-3-carboxylic acid, 97%
(R)-4-Boc-morpholine-3-carboxylic acid, 97%
Thermo Scientific Chemicals

(R)-4-Boc-morpholine-3-carboxylic acid, 97%

CAS: 869681-70-9 | C10H17NO5 | 231.248 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H64654.MD
also known as H64654-MD
Price (USD)
98.65
Online Exclusive
109.00
Save 10.35 (9%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
98.65
Online Exclusive
109.00
Save 10.35 (9%)
Each
Chemical Identifiers
CAS869681-70-9
IUPAC Name4-[(tert-butoxy)carbonyl]morpholine-3-carboxylic acid
Molecular FormulaC10H17NO5
InChI KeyKVXXEKIGMOEPSA-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)N1CCOCC1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Proton NMRConforms to structure
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥96.0% (NMR)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only