L-Serinamide hydrochloride, 95%
L-Serinamide hydrochloride, 95%
L-Serinamide hydrochloride, 95%
Thermo Scientific Chemicals

L-Serinamide hydrochloride, 95%

CAS: 65414-74-6 | C3H9ClN2O2 | 140.567 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H65169.06
also known as H65169-06
Price (USD)
69.65
Special offer
Online exclusive
Ends: 30-Jun-2026
82.00
Save 12.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
69.65
Special offer
Online exclusive
Ends: 30-Jun-2026
82.00
Save 12.35 (15%)
Each
Chemical Identifiers
CAS65414-74-6
IUPAC Namehydrogen 2-amino-3-hydroxypropanamide chloride
Molecular FormulaC3H9ClN2O2
InChI KeyVURWDDZIWBGXCK-UHFFFAOYNA-N
SMILES[H+].[Cl-].NC(CO)C(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥94.0%
FormPowder or crystals or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only