L-Glutamic acid 1-tert-butyl ester, 97%
L-Glutamic acid 1-tert-butyl ester, 97%
L-Glutamic acid 1-tert-butyl ester, 97%
Thermo Scientific Chemicals

L-Glutamic acid 1-tert-butyl ester, 97%

CAS: 45120-30-7 | C9H17NO4 | 203.238 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H65342.06
also known as H65342-06
Price (USD)
290.65
Online Exclusive
323.00
Save 32.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
290.65
Online Exclusive
323.00
Save 32.35 (10%)
Each
Chemical Identifiers
CAS45120-30-7
IUPAC Name4-amino-5-(tert-butoxy)-5-oxopentanoic acid
Molecular FormulaC9H17NO4
InChI KeyQVAQMUAKTNUNLN-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)C(N)CCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only