L-Alanine benzyl ester hydrochloride, 98%
L-Alanine benzyl ester hydrochloride, 98%
L-Alanine benzyl ester hydrochloride, 98%
Thermo Scientific Chemicals

L-Alanine benzyl ester hydrochloride, 98%

CAS: 5557-83-5 | C10H14ClNO2 | 215.677 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number H65699.22
also known as H65699-22
Price (USD)
378.65
Special offer
Online exclusive
Ends: 30-Jun-2026
445.00
Save 66.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
378.65
Special offer
Online exclusive
Ends: 30-Jun-2026
445.00
Save 66.35 (15%)
Each
Chemical Identifiers
CAS5557-83-5
IUPAC Namehydrogen benzyl 2-aminopropanoate chloride
Molecular FormulaC10H14ClNO2
InChI KeyRLMHWGDKMJIEHH-UHFFFAOYNA-N
SMILES[H+].[Cl-].CC(N)C(=O)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only