2-Cyclopropyl-L-glycine, 97%
2-Cyclopropyl-L-glycine, 97%
2-Cyclopropyl-L-glycine, 97%
Thermo Scientific Chemicals

2-Cyclopropyl-L-glycine, 97%

CAS: 49606-99-7 | C5H9NO2 | 115.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H65755.03
also known as H65755-03
Price (USD)
144.65
Online Exclusive
161.00
Save 16.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
144.65
Online Exclusive
161.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS49606-99-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
FormCrystals or powder or crystalline powder
Assay from Suppliers CofA≥96.0% (Titration)
Optical Rotation+97 to +115° (c=1, 1N HCl)
Proton NMRConforms to structure
2-Cyclopropyl-L-glycine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyclopropyl-L-glycine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water. Insoluble in ethanol.

Notes
Store at 4°C. Incompatible with heat and oxidizing agents. Protect from heat.
RUO – Research Use Only