N-Fmoc-L-glutamic acid 5-tert-butyl ester pentafluorophenyl ester, 98%
N-Fmoc-L-glutamic acid 5-tert-butyl ester pentafluorophenyl ester, 98%
N-Fmoc-L-glutamic acid 5-tert-butyl ester pentafluorophenyl ester, 98%
Thermo Scientific Chemicals

N-Fmoc-L-glutamic acid 5-tert-butyl ester pentafluorophenyl ester, 98%

CAS: 86061-04-3 | C30H26F5NO6 | 591.531 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H66008.14
also known as H66008-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Specifications
Beilstein3642680
CAS86061-04-3
Chemical Name or MaterialN-Fmoc-L-glutamic acid 5-tert-butyl ester pentafluorophenyl ester
Formula Weight591.53
IUPAC Name5-O-tert-butyl 1-O-(2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanedioate
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only