N-Boc-L-phenylalaninal, 97%, Thermo Scientific Chemicals
N-Boc-L-phenylalaninal, 97%, Thermo Scientific Chemicals
N-Boc-L-phenylalaninal, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-L-phenylalaninal, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H66530.03
also known as H66530-03
1 g
Catalog number H66530.03
also known as H66530-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS72155-45-4
IUPAC Nametert-butyl N-[(2S)-1-oxo-3-phenylpropan-2-yl]carbamate
Molecular FormulaC14H19NO3
InChI KeyZJTYRNPLVNMVPQ-LBPRGKRZSA-N
SMILESCC(C)(C)OC(=O)N[C@@H](CC1=CC=CC=C1)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
FormCrystals or powder or crystalline powder and/or chunks
Assay (GC)≥96.0%
Optical Rotation-41° to -49° (c=0.5, MeOH)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only