N-Fmoc-O-tert-butyl-L-allo-threonine, 97%
N-Fmoc-O-tert-butyl-L-allo-threonine, 97%
N-Fmoc-O-tert-butyl-L-allo-threonine, 97%
Thermo Scientific Chemicals

N-Fmoc-O-tert-butyl-L-allo-threonine, 97%

CAS: 201481-37-0 | C23H27NO5 | 397.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H66579.03
also known as H66579-03
Price (USD)
126.65
Special offer
Online exclusive
Ends: 30-Jun-2026
149.00
Save 22.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
126.65
Special offer
Online exclusive
Ends: 30-Jun-2026
149.00
Save 22.35 (15%)
Each
Chemical Identifiers
CAS201481-37-0
IUPAC Name(2S,3S)-3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Molecular FormulaC23H27NO5
InChI KeyLZOLWEQBVPVDPR-XOBRGWDASA-N
SMILESC[C@H](OC(C)(C)C)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only