N-Fmoc-4-benzoyl-L-phenylalanine, 95%
N-Fmoc-4-benzoyl-L-phenylalanine, 95%
N-Fmoc-4-benzoyl-L-phenylalanine, 95%
Thermo Scientific Chemicals

N-Fmoc-4-benzoyl-L-phenylalanine, 95%

CAS: 117666-96-3 | C31H25NO5 | 491.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H66707.MD
also known as H66707-MD
Price (USD)
59.65
Special offer
Online exclusive
Ends: 30-Jun-2026
70.20
Save 10.55 (15%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
59.65
Special offer
Online exclusive
Ends: 30-Jun-2026
70.20
Save 10.55 (15%)
Each
Chemical Identifiers
CAS117666-96-3
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only