N-Acetyl-L-tryptophan methyl ester, 95%
N-Acetyl-L-tryptophan methyl ester, 95%
N-Acetyl-L-tryptophan methyl ester, 95%
Thermo Scientific Chemicals

N-Acetyl-L-tryptophan methyl ester, 95%

CAS: 2824-57-9 | C14H16N2O3 | 260.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H66937.06
also known as H66937-06
Price (USD)
66.65
Special offer
Online exclusive
Ends: 30-Jun-2026
78.10
Save 11.45 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
66.65
Special offer
Online exclusive
Ends: 30-Jun-2026
78.10
Save 11.45 (15%)
Each
Specifications
CAS2824-57-9
Chemical Name or MaterialN-Acetyl-L-tryptophan methyl ester
ColorWhite
Formula Weight260.29
IUPAC Namemethyl (2S)-2-acetamido-3-(1H-indol-3-yl)propanoate
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only