CellInsight™ CX7 LED Pro High Content Screening Platform
CellInsight™ CX7 LED Pro High Content Screening Platform
CellInsight™ CX7 LED Pro High Content Screening Platform
Thermo Scientific™
CellInsight™ CX7 LED Pro High Content Screening Platform
The Thermo Scientific CellInsight CX7 LED Pro High Content Screening (HCS) Platform is a fast, LED engine-based, automated cellular imagingRead more
Have Questions?
Catalog NumberQuantity
HCSDCX7LEDPRO1 each
Catalog number HCSDCX7LEDPRO
Price (MXN)
-
Quantity:
1 each
The Thermo Scientific CellInsight CX7 LED Pro High Content Screening (HCS) Platform is a fast, LED engine-based, automated cellular imaging and analysis platform designed for life scientists whom require on-the-fly quantitative analysis. It offers an integrated way to develop and automate high content assays using a combination of 5-color brightfield and 7-color fluorescence imaging in confocal or wide-field modes.
Features of the CellInsight CX7 LED Pro Platform
• Back-illuminated sCMOS camera offer a 95% peak quantum efficiency and -20°C cooling
• New validated applications including Cell Painting and FirePlex-HT immunoassay
• Olympus X-line objective imaging to improve imaging quality and assay performance
• EurekaScan Finder double-pass functionality for on-the-fly seek and find of spheroids, tissue, and rare cellular events
• HCS Studio 5.0 with over 40 life science bio-applications and real-time analysis for fastest time-to-data possible
• 7-channel fluorescent imaging
• 5-channel bright-field imaging
• 7-channel confocal imaging
• Software and laser-based autofocus for consistent scan times
• Configured for fully automated plate handling and scanning
Capabilities
• Use HCS Studio software for integrated instrument control and data analysis
• Achieve the fastest time to results by using intelligent acQuisition (iQ) real-time analysis feedback
• Addresses hundreds of assay types with flexible software and hardware tools
• Scan and analyze microscope slides or most SBS-standard format microplates
• Compatible with Molecular Probes fluorescent detection reagents
Hardware and software
• Stable and reliable light source enables reduced variability, increased data quality, and decreased operating costs
• Broad reagent options with 7-color excitation from violet to near IR for assay multiplexing
• Dual mode focusing using Software and Laser Autofocus methods to address both labelled and unlabeled sample types
• Multi-mode imaging allows for wide-field, bright-field, and confocal imaging in one run
• HCS Studio 5.0 Cell Analysis Software is an integrated instrument control, visualization, and analysis software package for novice to advanced users
• Scalable with Store Image and Database Software and optional Orbitor RS Microplate handler (not included)
For Research Use Only.
Specifications
Quantity1 each
ShakingLinear, Orbital, Double Orbital
Voltage0 to 3000 VDC, 0 to 250 VAC
CameraHighly Sensitive Monochrome CMOS
Product LineCellInsight
Unit SizeEach