Human Hematopoietic Stem Cell Expansion Cytokine Package (IL-3), PeproTech®
Gibco™
Human Hematopoietic Stem Cell Expansion Cytokine Package (IL-3), PeproTech®
The Human Hematopoietic Stem Cell Expansion Cytokine Package contains the key components required for ex vivo human hematopoietic stem cells.
Have Questions?
Catalog NumberQuantity
HHSC31 Ea.
Catalog number HHSC3
Price (USD)
1,672.65
Online exclusive
2,230.00
Save 557.35
Each
Quantity:
1 Ea.
Price (USD)
1,672.65
Online exclusive
2,230.00
Save 557.35
Each
The Human Hematopoietic Stem Cell Expansion Cytokine Package contains recombinant Flt3 ligand, stem cell factor (SCF), thrombopoietin (TPO), and interleukin-3 (IL-3), critical growth factors and cytokines used in ex vivo culture of hematopoietic stem cells (HSCs). These factors support the proliferation, survival, and differentiation of HSCs.
The package contains the following four vials:
  • 100 μg Human Flt3 Ligand (Cat. No. 300-19-100UG)
  • 100 μg Human SCF (Cat. No. 300-07-100UG)
  • 100 μg Human TPO (Cat. No. 300-18-100UG)
  • 10 μg Human IL-3 (Cat. No. 200-03-10UG)
Flt3 ligand is a growth factor that regulates proliferation of early hematopoietic cells. By itself it does not stimulate proliferation of early hematopoietic cells, but synergizes with other CSFs and interleukins to induce growth and differentiation.
Human SCF is a hematopoietic growth factor that exerts its activity by signaling through the c-Kit receptor. Human SCF and c-Kit are essential for the survival, proliferation, and differentiation of hematopoietic stem cells committed to the melanocyte and germ cell lineages. Human SCF manifests low activity on murine cells, while murine and rat SCF are fully active on human cells.
TPO is a lineage-specific growth factor produced in the liver, kidney, and skeletal muscle. It stimulates the proliferation and maturation of megakaryocytes from hematopoietic stem cells and promotes increased circulating levels of platelets in vivo.
IL-3 is a hematopoietic growth factor that promotes the survival, differentiation, and proliferation of committed progenitor cells of the megakaryocyte, granulocyte-macrophage, erythroid, eosinophil, basophil, and mast cell lineages.
Specifications
For Use With (Application)Functional Assay
FormulationLyophilized
Quantity1 Ea.
SpeciesHuman
Product TypeStem Cell Expansion Cytokine Package
Unit SizeEach
Frequently asked questions (FAQs)
Which cytokines show cross-species activity?
With a few exceptions, most human cytokines are active on mouse cells. Many mouse cytokines are active on human cells, but may show lower specific activity than the corresponding human cytokine. The interferons, GM-CSF, IL-3, and IL-4 are known to be species-specific with very little, if any, activity on non-homologous cells. In contrast, the FGFs and neurotrophins are very highly conserved and show excellent activity on cells of other animal species.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.