Human Hematopoietic Stem Cell Expansion Cytokine Package (IL-6), PeproTech®
Gibco™
Human Hematopoietic Stem Cell Expansion Cytokine Package (IL-6), PeproTech®
The Human Hematopoietic Stem Cell Expansion Cytokine Package contains the key components required for ex vivo human hematopoietic stem cells.
Have Questions?
Catalog NumberQuantity
HHSC61 Pack
Catalog number HHSC6
Price (USD)
1,672.65
Online exclusive
2,230.00
Save 557.35
Each
Quantity:
1 Pack
Price (USD)
1,672.65
Online exclusive
2,230.00
Save 557.35
Each
The Human Hematopoietic Stem Cell Expansion Cytokine Package contains recombinant Flt3 ligand, stem cell factor (SCF), thrombopoietin (TPO), and interleukin-6 (IL-6), critical growth factors and cytokines use in ex vivo culture of hematopoietic stem cells (HSCs). These factors support the proliferation, survival, and differentiation of HSCs.
The package contains the following 4 vials:
  • 100 μg Human Flt3 Ligand (Cat. No. 300-19-100UG)
  • 100 μg Human SCF (Cat. No. 300-07-100UG)
  • 100 μg Human TPO (Cat. No. 300-18-100UG)
  • 20 μg Human IL-6 (Cat. No. 200-06-20UG)
Flt3 ligand is a growth factor that regulates proliferation of early hematopoietic cells. By itself it does not stimulate proliferation of early hematopoietic cells, but synergizes with other CSFs and interleukins to induce growth and differentiation.
SCF is a hematopoietic growth factor that exerts its activity by signaling through the c-Kit receptor. SCF and c-Kit are essential for the survival, proliferation, and differentiation of hematopoietic cells committed to the melanocyte and germ cell lineages. Human SCF manifests low activity on murine cells, while murine and rat SCF are fully active on human cells.
TPO is a lineage-specific growth factor produced in the liver, kidney, and skeletal muscle. It stimulates the proliferation and maturation of megakaryocytes and promotes increased circulating levels of platelets in vivo.
IL-6 is a pleiotropic cytokine that stimulates B-cell differentiation and antibody production, synergizes with IL-3 in megakaryocyte development and platelet production, induces expression of hepatic acute-phase proteins, and regulates bone metabolism. IL-6 signals through the IL-6 receptor system that consists of two chains, IL-6R and gp130. Recombinant Human IL-6 is a 20.9 kDa protein containing 184 amino acid residues.
Specifications
For Use With (Application)Functional Assay
FormulationLyophilized
Quantity1 Pack
SpeciesHuman
Product TypeStem Cell Expansion Cytokine Package
Unit SizeEach
Frequently asked questions (FAQs)
Which cytokines show cross-species activity?
With a few exceptions, most human cytokines are active on mouse cells. Many mouse cytokines are active on human cells, but may show lower specific activity than the corresponding human cytokine. The interferons, GM-CSF, IL-3, and IL-4 are known to be species-specific with very little, if any, activity on non-homologous cells. In contrast, the FGFs and neurotrophins are very highly conserved and show excellent activity on cells of other animal species.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.