Will I be able to transfect my primary hepatocyte cells if I use an overlay?
No. The overlay will interfere with transfection. For transfection assays, we recommend that you perform the transfection first, and then proceed to overlay the cells.
Search
View additional product information for Human Suspension Hepatocytes, Polymorphic Donors, (4-8 x 106 cells) - FAQs (HMCSPS)
1 product FAQs found