Iodoacetamide Alkyne
Invitrogen™

Iodoacetamide Alkyne

Iodoacetamide Alkyne
Have Questions?
Catalog NumberQuantity
I101891 mg
Catalog number I10189
Price (USD)
449.00
Each
Quantity:
1 mg
Price (USD)
449.00
Each

Conjugates prepared with the thiol-reactive alkyne, succinimidyl ester can be detected with an azide-containing molecule in a click chemistry reaction. Click chemistry describes a class of chemical reactions that use bio-orthogonal or biologically unique moities to label and detect a molecule of interest using a two-step procedure. The two-step reaction procedure involves a copper-catalyzed triazole formation of an azide and an alkyne. Click reactions have several characteristics: the reaction between the detection moieties is efficient; no extreme temperatures or solvents are required; the reaction product is stable; the components of the reaction are bioinert; and perhaps most importantly, no side reactions occur – the label and detection tags react selectively and specifically with one another. Unlike traditional chemical reactions utilizing succinimidyl esters or maleimides that target amines and sulfhydryls – functional groups that are not unique – click chemistry-labeled molecules can be applied to complex biological samples and be detected with unprecedented sensitivity due to extremely low background.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Chemical ReactivityThiol
FormatSolid
Label or DyeAlkyne
Molecular Weight (g/mol)223.01 Da
Product TypeIodoacetamide Alkyne
Quantity1 mg
Reactive GroupIodoacetamide
Reactive MoietyAlkyl Halide, Iodoacetamide
Shipping ConditionRoom Temperature
SolubilityDMSO (Dimethylsulfoxide)
Label TypeClick Chemistry Label
Product LineMolecular Probes
Unit SizeEach
Contents & Storage
Store at ≤-20°C, desiccated and protect from light.

Citations & References (1)

Citations & References
Abstract
Reversibility of covalent electrophile-protein adducts and chemical toxicity.
Authors:Lin D, Saleh S, Liebler DC,
Journal:Chem Res Toxicol
PubMed ID:19548357
The biotin-tagged electrophiles 1-biotinamido-4-(4'-[maleimidoethylcyclohexane]-carboxamido)butane (BMCC) and N-iodoacetyl-N-biotinylhexylenediamine (IAB) have been used as model electrophile probes in complex proteomes to identify protein targets associated with chemical toxicity. Whereas IAB activates stress signaling and apoptosis in HEK293 cells, BMCC does not. Cysteine Michael adducts formed from BMCC and nonbiotinylated analogues rapidly disappeared ... More