Isolectin GS-IB<sub>4</sub> From <i>Griffonia simplicifolia</i>, Alexa Fluor&trade; 594 Conjugate
Invitrogen™

Isolectin GS-IB4 From Griffonia simplicifolia, Alexa Fluor™ 594 Conjugate

Isolectin GS-IB4 is a 114,000-dalton glycoprotein that is part of a family of five tetrameric type I isolectins (IA4, IA3B,Read more
Have Questions?
Catalog NumberQuantity
I21413500 μg
Catalog number I21413
Price (USD)
475.60
Special offer
Online exclusive
Ends: 31-Dec-2026
580.00
Save 104.40
Each
Quantity:
500 μg
Price (USD)
475.60
Special offer
Online exclusive
Ends: 31-Dec-2026
580.00
Save 104.40
Each
Isolectin GS-IB4 is a 114,000-dalton glycoprotein that is part of a family of five tetrameric type I isolectins (IA4, IA3B, IA2B2, IAB3, and IB4) isolated from the seeds of the tropical African legume Griffonia simplicifolia. The A subunit prefers N-acetyl-D-galactosamine end groups while the B subunit is selective for terminal a-D-galactosyl residues. The red fluorescent Alexa Fluor™ 594 isolectin GS-IB4 conjugate can be used in fluorescence microscopy.

View complete list of fluorescent dye-conjugated lectins ›
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label TypeAlexa Fluor Dyes
Product LineAlexa Fluor
Protein SubtypeLectins
Quantity500 μg
Shipping ConditionRoom Temperature
SourceAfrica Shrub legume
ConjugateAlexa Fluor 594
FormLyophilized
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Frequently asked questions (FAQs)

How do I know which tracer to choose for my experiment?

Factors to consider are size of tracer, method of delivery (injection, direct application to tissue, etc.), and if the tracer needs to be fixable. Here are some links to details about the various classes of neuronal tracers we offer and how to choose between them:

Neuronal Tracing (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-tracing-tracking-and-morphology/neuronal-tracing.html)
Choosing a Tracer (https://www.thermofisher.com/us/en/home/references/molecular-probes-the-handbook/fluorescent-tracers-of-cell-morphology-and-fluid-flow/choosing-a-tracer.html)
Imaging Analysis (http://assets.thermofisher.com/TFS-Assets/BID/Reference-Materials/bioprobes-50-journal.pdf)

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (18)

Citations & References
Abstract
Insulin-like growth factor 2 and potential regulators of hemangioma growth and involution identified by large-scale expression analysis.
Authors:Ritter MR, Dorrell MI, Edmonds J, Friedlander SF, Friedlander M
Journal:Proc Natl Acad Sci U S A
PubMed ID:12032304
'Hemangiomas are benign tumors of the vascular endothelium and are the most common tumors of infancy. These tumors are characterized by an initial phase of rapid proliferation, which is followed, in most cases, by spontaneous involution. Although most lesions resolve without complication, there are some cases in which hemangiomas can ... More
Cell fusion-independent differentiation of neural stem cells to the endothelial lineage.
Authors:Wurmser AE, Nakashima K, Summers RG, Toni N, D'Amour KA, Lie DC, Gage FH
Journal:Nature
PubMed ID:15254537
'Somatic stem cells have been claimed to possess an unexpectedly broad differentiation potential (referred to here as plasticity) that could be induced by exposing stem cells to the extracellular developmental signals of other lineages in mixed-cell cultures. Recently, this and other experimental evidence supporting the existence of stem-cell plasticity have ... More
Effects of familial hemiplegic migraine type 1 mutation T666M on voltage-gated calcium channel activities in trigeminal ganglion neurons.
Authors:Tao J, Liu P, Xiao Z, Zhao H, Gerber BR, Cao YQ,
Journal:J Neurophysiol
PubMed ID:22190617
'Familial hemiplegic migraine type 1 (FHM-1), a rare hereditary form of migraine with aura and hemiparesis, serves as a good model for exploring migraine pathophysiology. The FHM-1 gene encodes the pore-forming Ca(V)2.1 subunit of human P/Q-type voltage-gated Ca(2+) channels (VGCCs). Some FHM-1 mutations result in a decrease of whole cell ... More
CXCL16 orchestrates adenosine A3 receptor and MCP-1/CCL2 activity to protect neurons from excitotoxic cell death in the CNS.
Authors:Rosito M, Deflorio C, Limatola C, Trettel F,
Journal:J Neurosci
PubMed ID:22378888
A role for chemokines as molecules mediating neuron-glia cross talk has emerged in recent years, both in physiological and pathological conditions. We demonstrate here for the first time that the chemokine CXCL16 and its unique receptor CXCR6 are functionally expressed in the CNS, and induce neuroprotection against excitotoxic damage due ... More
Microglial cells in astroglial cultures: a cautionary note.
Authors:Saura J,
Journal:J Neuroinflammation
PubMed ID:17937799
Primary rodent astroglial-enriched cultures are the most popular model to study astroglial biology in vitro. From the original methods described in the 1970's a great number of minor modifications have been incorporated into these protocols by different laboratories. These protocols result in cultures in which the astrocyte is the predominant ... More