Isolectin GS-IB4 From Griffonia simplicifolia, Alexa Fluor™ 594 Conjugate - FAQs
Isolectin GS-IB4 From Griffonia simplicifolia, Alexa Fluor™ 594 Conjugate - FAQs
View additional product information for Isolectin GS-IB4 From Griffonia simplicifolia, Alexa Fluor™ 594 Conjugate - FAQs (I21413)
1 product FAQs found
How do I know which tracer to choose for my experiment?
Factors to consider are size of tracer, method of delivery (injection, direct application to tissue, etc.), and if the tracer needs to be fixable. Here are some links to details about the various classes of neuronal tracers we offer and how to choose between them:
Neuronal Tracing (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-tracing-tracking-and-morphology/neuronal-tracing.html)
Choosing a Tracer (https://www.thermofisher.com/us/en/home/references/molecular-probes-the-handbook/fluorescent-tracers-of-cell-morphology-and-fluid-flow/choosing-a-tracer.html)
Imaging Analysis (http://assets.thermofisher.com/TFS-Assets/BID/Reference-Materials/bioprobes-50-journal.pdf)