Isolectin GS-IB4 From Griffonia simplicifolia, Alexa Fluor™ 647 Conjugate
Isolectin GS-IB4 is a 114,000-dalton glycoprotein that is part of a family of five tetrameric type I isolectins (IA4, IA3B,Read more
Have Questions?
Catalog Number
Quantity
I32450
500 μg
Catalog number I32450
Price (USD)
541.65
Online exclusive
582.00
Save 40.35
Each
Quantity:
500 μg
Price (USD)
541.65
Online exclusive
582.00
Save 40.35
Each
Isolectin GS-IB4 is a 114,000-dalton glycoprotein that is part of a family of five tetrameric type I isolectins (IA4, IA3B, IA2B2, IAB3, and IB4) isolated from the seeds of the tropical African legume Griffonia simplicifolia. The A subunit prefers N-acetyl-D-galactosamine end groups while the B subunit is selective for terminal a-D-galactosyl residues. The far-red fluorescent Alexa Fluor™ 647 isolectin GS-IB4 conjugate can be used in fluorescence microscopy or flow cytometry.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label TypeAlexa Fluor Dyes
Product LineAlexa Fluor
Protein SubtypeLectins
Quantity500 μg
Shipping ConditionRoom Temperature
SourceAfrica Shrub legume
ConjugateAlexa Fluor 647
FormLyophilized
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.
Citations & References (17)
Citations & References
Abstract
Cell fusion-independent differentiation of neural stem cells to the endothelial lineage.
Authors:Wurmser AE, Nakashima K, Summers RG, Toni N, D'Amour KA, Lie DC, Gage FH
Journal:Nature
PubMed ID:15254537
'Somatic stem cells have been claimed to possess an unexpectedly broad differentiation potential (referred to here as plasticity) that could be induced by exposing stem cells to the extracellular developmental signals of other lineages in mixed-cell cultures. Recently, this and other experimental evidence supporting the existence of stem-cell plasticity have ... More
Early activation, motility, and homing of neonatal microglia to injured neurons does not require protein synthesis.
Authors:Kurpius D, Wilson N, Fuller L, Hoffman A, Dailey ME
Journal:Glia
PubMed ID:16715500
'Neuronal injury in CNS tissues induces a rapid activation and mobilization of resident microglia (MG). It is widely assumed that changes in gene expression drive the morphological transformation of MG and regulate their mobilization during activation. Here, we used acutely excised neonatal rat brain slices to test whether the morphological ... More
Phenotypic characterization of gastric sensory neurons in mice.
Authors:Bielefeldt K, Zhong F, Koerber HR, Davis BM,
Journal:Am J Physiol Gastrointest Liver Physiol
PubMed ID:16728726
Recent studies suggest that the capsaicin receptor [transient receptor potential vanilloid (TRPV)1] may play a role in visceral mechanosensation. To address the potential role of TRPV1 in vagal sensory neurons, we developed a new in vitro technique allowing us to determine TRPV1 expression directly in physiologically characterized gastric sensory neurons. ... More
Sequences in intron 51 of the von Willebrand factor gene target promoter activation to a subset of lung endothelial cells in transgenic mice.
Authors:Kleinschmidt AM, Nassiri M, Stitt MS, Wasserloos K, Watkins SC, Pitt BR, Jahroudi N,
Journal:J Biol Chem
PubMed ID:18048367
In vivo analyses of the VWF promoter previously demonstrated that a fragment spanning sequences -487 to +247 targets promoter activation to brain vascular endothelial cells, whereas a longer fragment including 2182 bp of the 5'-flanking sequences, the first exon, and the first intron activated expression in endothelial cells of the ... More
A new method to isolate microglia from adult mice and culture them for an extended period of time.
Authors:Moussaud S, Draheim HJ,
Journal:J Neurosci Methods
PubMed ID:20097228
As the major immuno-competent cells of the brain, microglia are highly implicated in neuro-protection as well as in neurodegeneration. Therefore, they are of key interest for research on numerous CNS diseases. Currently, to model inflammation in the brain, microglial cell lines or primary microglia prepared from embryonic or neo-natal rodents ... More