INTELLIFLEX Calibration Kit
Invitrogen™
INTELLIFLEX Calibration Kit
The INTELLIFLEX Calibration Kit is for use with the Luminex xMAP INTELLIFLEX and Luminex xMAP INTELLIFLEX DR-SE systems and is used to calibrate the optics of these systems.
Have Questions?
Catalog NumberQuantity
IFXCALK2020 reactions kit
Catalog number IFXCALK20
Price (USD)
660.00
Each
Quantity:
20 reactions kit
Price (USD)
660.00
Each
The INTELLIFLEX Calibration Kit is for use with the Luminex xMAP INTELLIFLEX and Luminex xMAP INTELLIFLEX DR-SE systems and is used to calibrate the optics of these systems. The calibration kit contains all reagents needed for calibration of these systems.
Calibration is important to ensure that the optical system functions effectively and that different INTELLIFLEX systems report similar results. Calibrating the INTELLIFLEX System normalizes the settings for classification channels (CL1, CL2, and CL3), the doublet discriminator channel (DD), and the reporter channels (RP1 and RP2).
Please refer to the INTELLIFLEX Calibration Kit package insert for proper use of the INTELLIFLEX Calibration Kit.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Luminex™ xMAP™ INTELLIFLEX, Luminex™ xMAP™ INTELLIFLEX DR-SE
No. of Tests20 Calibrations
Quantity20 reactions kit
Shipping ConditionWet Ice
Unit SizeEach
Contents & Storage
• xMAP INTELLIFLEX Calibrator, 10 mL
• 25 Strips wells
Each kit will provide 20 calibrations.
Store in the dark at 2°C to 8°C.