TRAM 34
TRAM 34
TRAM 34
Thermo Scientific Chemicals

TRAM 34

CAS: 289905-88-0 | C22H17ClN2, C22H17ClN2 | 344.84 g/mol
Have Questions?
Catalog NumberQuantity
J60019.MB
also known as J60019-MB
25 mg
Catalog number J60019.MB
also known as J60019-MB
Price (USD)
271.65
Online exclusive
302.00
Save 30.35
Each
Quantity:
25 mg
Request bulk or custom format
Price (USD)
271.65
Online exclusive
302.00
Save 30.35
Each
Chemical Identifiers
CAS289905-88-0
IUPAC Name1-[(2-chlorophenyl)diphenylmethyl]-1H-pyrazole
Molecular FormulaC22H17ClN2
InChI KeyKBFUQFVFYYBHBT-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1C(N1C=CC=N1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Purity≥95.0%
Proton NMRConforms
TRAM-34 blocks intermediate conductance calcium-activated potassium channel IKCa1 with a Kd of 20nM and exhibits exquisite selectivity for the channel. Clotrimazole impurity. Also used to study its effect on the prevention of vascular restenosis after balloon angioplasty.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
2°C to 8°C